missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-1-methyl-1H-pyrazole, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-iodo-1-methyl-1h-pyrazole, 1-methyl-3-iodopyrazole, 1h-pyrazole, 3-iodo-1-methyl, 3-iodo-1-methy-1h-pyrazole, iodo-n-methylpyrazole, 3-idio-1-methylpyrazole, 3-iodanyl-1-methyl-pyrazole, 1h-pyrazole,3-iodo-1-methyl, 3 5-iodopyrazole, 1-me |
Formula Weight | 208 |
Percent Purity | 97% |
Chemical Name or Material | 3-Iodo-1-methyl-1H-pyrazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'