missing translation for 'onlineSavingsMsg'
Learn More

3-Indolebutyric acid, 98%, Thermo Scientific Chemicals

Catalog Number p-4112282
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
133-32-4
C12H13NO2
203.24
MFCD00005664
JTEDVYBZBROSJT-UHFFFAOYSA-N
indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
8617
CHEBI:33070
4-(1H-indol-3-yl)butanoic acid
C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O
This item is not returnable. View return policy
133-32-4
C12H13NO2
203.24
MFCD00005664
JTEDVYBZBROSJT-UHFFFAOYSA-N
indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
8617
CHEBI:33070
4-(1H-indol-3-yl)butanoic acid
C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 120.0°C to 125.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 22, II, 54
Merck Index 15, 5005
Solubility Information Solubility in water: 250mg/l (20°C). Other solubilities: soluble in ethanol,ether,acetone,benzene,,dimethyl sulfoxide and petroleum ether,insoluble in chloroform,5% in ethanol: clear,brownish solution
Formula Weight 203.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Indolebutyric acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Indolebutyric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'