missing translation for 'onlineSavingsMsg'
Learn More

3-Indolebutyric acid, 98%, ACROS Organics™

Brand:  Acros Organics 160691000

Code : 29

Additional Details : CAS Number : 133-32-4 Weight : 0.15000kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10523104

  • £205.90 / 100g


EU Stock 4
Add to basket


Specifications

Chemical Identifiers

3-Indolebutyric acid
97.5
98%
C12H13NO2
MFCD00005664
133-32-4
100.0
98%
203.24
4-(3-1H-Indolyl)butyric acid

Specifications

3-Indolebutyric acid
97.5
10ppm max.
Glass bottle
98%
C12H13NO2
MFCD00005664
22, II, 54
15, 5005
120.0° to 125.0°C
100g
133-32-4
100.0
Authentic
Liquid
98%
203.24
0.1% max.
0.5% max. (K.F.)
4-(3-1H-Indolyl)butyric acid
Clear colorless to pale yellow
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Indolebutyric acid

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS