missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyquinoline, +97%, ACROS Organics™

Brand:  Acros Organics 391372500

Code : 29

Additional Details : CAS Number : 580-18-7 Weight : 0.05025kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10482975

  • £84.90 / 250mg


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

3-Hydroxyquinoline
97.0
≥97.0%
C9H7NO
MFCD00169018
580-18-7
100.0
97+%
145.16
3-Quinolinol

Specifications

3-Hydroxyquinoline
198° to 202°C
580-18-7
100.0
Glass bottle
≥97.0%
C9H7NO
MFCD00169018
Beige
250mg
97.0
Authentic
Crystalline Powder
97+%
145.16
3-Quinolinol
Videos
SDS