missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (±)-3-Hydroxypyrrolidine hydrochloride, 98%

Brand:  Alfa Aesar™ H64975.14

Code : NEW

Additional Details : CAS Number : 86070-82-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15402419

  • £85.19 / 25g

Estimated Shipment: 19-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

86070-82-8
123.58
QPMSJEFZULFYTB-UHFFFAOYNA-N
2769408
Cl.OC1CCNC1
C4H10ClNO
MFCD03001754
pyrrolidin-3-ol hydrochloride, 3-hydroxypyrrolidine hydrochloride, 3-pyrrolidinol hydrochloride, 3-hydroxypyrrolidine hcl, 3-pyrrolidinol hcl, 3-pyrrolidinol, hydrochloride, 3-hydroxypyrrolidinehydrochloride, s-3-hydroxypyrrolidine hcl, 3-pyrrolidinol hci, hydroxypyrrolidine hcl
pyrrolidin-3-ol;hydrochloride

Specifications

(±)-3-Hydroxypyrrolidine hydrochloride
86070-82-8
MFCD03001754
pyrrolidin-3-ol hydrochloride, 3-hydroxypyrrolidine hydrochloride, 3-pyrrolidinol hydrochloride, 3-hydroxypyrrolidine hcl, 3-pyrrolidinol hcl, 3-pyrrolidinol, hydrochloride, 3-hydroxypyrrolidinehydrochloride, s-3-hydroxypyrrolidine hcl, 3-pyrrolidinol hci, hydroxypyrrolidine hcl
Cl.OC1CCNC1
123.58
123.58
98%
25g
C4H10ClNO
7029051
QPMSJEFZULFYTB-UHFFFAOYNA-N
pyrrolidin-3-ol;hydrochloride
2769408
Solid
Videos
SDS Documents
Certificates