missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Hydroxypyrazine-2-carboxamide, 98%

£18.70 - £168.32

Chemical Identifiers

CAS 55321-99-8
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.114
MDL Number MFCD00233977
InChI Key SZPBAPFUXAADQV-UHFFFAOYSA-N
Synonym 3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry
PubChem CID 294642
IUPAC Name 2-oxo-1H-pyrazine-3-carboxamide
SMILES C1=CN=C(C(=O)N1)C(=O)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15453817
SDS View Documents
Alfa Aesar™
H28250.MD
250mg £18.70
250mg View alternative products
Save Up to 
»
In stock, ready to ship: 2 Please to purchase this item. Need a web account? Register with us today!
11387639
SDS View Documents
Alfa Aesar™
H28250.03
1g £51.12
1g View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
11397639
SDS View Documents
Alfa Aesar™
H28250.06
5g £168.32
5g View alternative products
Save Up to 
»
Estimated Shipment
10-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications
Apart from biological importance of pyridine-2-carboxamide (picolinamide, pia) as well as of its derivatives such as 3-hydroxypyridine-2-carboxamide (3-OHpia), as ligands they have found their extensive application in coordina- tion chemistry in crystal engineering .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

55321-99-8
139.114
SZPBAPFUXAADQV-UHFFFAOYSA-N
294642
C1=CN=C(C(=O)N1)C(=O)N
C5H5N3O2
MFCD00233977
3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry
2-oxo-1H-pyrazine-3-carboxamide

Specifications

3-Hydroxypyrazine-2-carboxamide
250mg
C5H5N3O2
3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry
C1=CN=C(C(=O)N1)C(=O)N
139.114
139.12
∽270°C (decomposition)
55321-99-8
MFCD00233977
SZPBAPFUXAADQV-UHFFFAOYSA-N
2-oxo-1H-pyrazine-3-carboxamide
294642
98%