missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypyrazine-2-carboxamide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Apart from biological importance of pyridine-2-carboxamide (picolinamide, pia) as well as of its derivatives such as 3-hydroxypyridine-2-carboxamide (3-OHpia), as ligands they have found their extensive application in coordina- tion chemistry in crystal engineering .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Apart from biological importance of pyridine-2-carboxamide (picolinamide, pia) as well as of its derivatives such as 3-hydroxypyridine-2-carboxamide (3-OHpia), as ligands they have found their extensive application in coordina- tion chemistry in crystal engineering .
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Apart from biological importance of pyridine-2-carboxamide (picolinamide, pia) as well as of its derivatives such as 3-hydroxypyridine-2-carboxamide (3-OHpia), as ligands they have found their extensive application in coordina- tion chemistry in crystal engineering .
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | ∽270°C (decomposition) |
Quantity | 1 g |
Formula Weight | 139.12 |
Percent Purity | 98% |
Chemical Name or Material | 3-Hydroxypyrazine-2-carboxamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'