missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypiperidine hydrochloride, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 196.6°C to 198.7°C |
Color | White |
Elemental Analysis | 42.97 to 44.28% C, 9.90 to 10.33% N |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Quantity | 5 g |
Formula Weight | 137.61 |
Percent Purity | 99% |
Show More |
Safety and Handling
Product Identifier
- 3-Hydroxypiperidine hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'