missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxymethyl-4-methoxybenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7056004
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
908142-03-0
C8H11BO4
181.982
MFCD14155851
DOQUCBSZQOPLGT-UHFFFAOYSA-N
3-hydroxymethyl-4-methoxyphenyl boronic acid, 3-hydroxymethyl-4-methoxyphenylboronic acid, ksc005q4b, 3-hydroxymethyl-4-methoxybenzeneboronic acid, 3-hydroxymethyl-4-methoxyphenyl boronicacid, b-3-hydroxymethyl-4-methoxyphenyl-boronic acid, boronic acid, b-3-hydroxymethyl-4-methoxyphenyl
53407782
[3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
B(C1=CC(=C(C=C1)OC)CO)(O)O
This item is not returnable. View return policy
908142-03-0
C8H11BO4
181.982
MFCD14155851
DOQUCBSZQOPLGT-UHFFFAOYSA-N
3-hydroxymethyl-4-methoxyphenyl boronic acid, 3-hydroxymethyl-4-methoxyphenylboronic acid, ksc005q4b, 3-hydroxymethyl-4-methoxybenzeneboronic acid, 3-hydroxymethyl-4-methoxyphenyl boronicacid, b-3-hydroxymethyl-4-methoxyphenyl-boronic acid, boronic acid, b-3-hydroxymethyl-4-methoxyphenyl
53407782
[3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
B(C1=CC(=C(C=C1)OC)CO)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Beilstein 15501236
Solubility Information Slightly soluble in water.
Formula Weight 181.98
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Hydroxymethyl-4-methoxybenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'