missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxybutyric acid, tech., Thermo Scientific Chemicals
51.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Industrially, 3-hydroxybutanoic acid is used for the manufacture of biodegradable plastics. It is also acts as a solvent in organoleptics, cosmetics. It is also used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by Escherichia coli.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 1.13 g/mL |
Boiling Point | 130°C (12 mmHg) |
Flash Point | >112°C (234°F) |
Refractive Index | 1.4445 |
Quantity | 5 g |
Sensitivity | Hygroscopic |
Merck Index | 14,4816 |
Solubility Information | Soluble in water, alcohol, ether |
Formula Weight | 104.1 |
Show More |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'