missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybutyric acid, 95%, Thermo Scientific Chemicals

Catalog Number p-3778777
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
300-85-6
C4H8O3
104.11
MFCD00004546
WHBMMWSBFZVSSR-UHFFFAOYSA-N
3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy
441
CHEBI:20067
3-hydroxybutanoic acid
CC(CC(=O)O)O
This item is not returnable. View return policy
300-85-6
C4H8O3
104.11
MFCD00004546
WHBMMWSBFZVSSR-UHFFFAOYSA-N
3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy
441
CHEBI:20067
3-hydroxybutanoic acid
CC(CC(=O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Density 1.1260g/mL
Boiling Point 118.0°C to 120.0°C (2.0 mmHg)
Flash Point 112°C
Assay Percent Range 95&percent;
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2CO2H
Refractive Index 1.4420 to 1.4440
Quantity 5 g
Beilstein 03, 307
Fieser 14,178
Merck Index 15, 4855
Specific Gravity 1.126
Solubility Information Solubility in water: >10% in water. Other solubilities: soluble in alcohol,ether
Formula Weight 104.11
Percent Purity 95%
Grade Technical
Chemical Name or Material 3-Hydroxybutyric acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxybutyric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'