missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-6-methylpyridine, 99%, Thermo Scientific Chemicals

Catalog Number 10591771
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1121-78-4
C6H7NO
109.13
MFCD00006339
DHLUJPLHLZJUBW-UHFFFAOYSA-N
5-hydroxy-2-methylpyridine, 2-methyl-5-hydroxypyridine, 3-hydroxy-6-methylpyridine, 6-methyl-3-pyridinol, 3-pyridinol, 6-methyl, 6-methyl-3-hydroxypyridine, 5-hydroxy-2-picoline, unii-uke16jp1jr, 2-methyl-5-pyridinol, uke16jp1jr
14275
6-methylpyridin-3-ol
CC1=NC=C(C=C1)O
This item is not returnable. View return policy
1121-78-4
C6H7NO
109.13
MFCD00006339
DHLUJPLHLZJUBW-UHFFFAOYSA-N
5-hydroxy-2-methylpyridine, 2-methyl-5-hydroxypyridine, 3-hydroxy-6-methylpyridine, 6-methyl-3-pyridinol, 3-pyridinol, 6-methyl, 6-methyl-3-hydroxypyridine, 5-hydroxy-2-picoline, unii-uke16jp1jr, 2-methyl-5-pyridinol, uke16jp1jr
14275
6-methylpyridin-3-ol
CC1=NC=C(C=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 168°C to 171°C
Color Beige to Tan
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C5H4N)CH2OH
Quantity 100 g
Beilstein 21, III, 480
Formula Weight 109.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-6-methylpyridine, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'