missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-nitrobenzoic Acid 98%, ACROS Organics™

Brand:  Acros Organics 121671000

Code : 29

Additional Details : CAS Number : 619-14-7 Weight : 0.15000kg


Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10644972

  • £131.40 / 100g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

3-Hydroxy-4-nitrobenzoic acid
619-14-7
100.0
98%
HOC6H3(NO2)CO2H
MFCD00007110
98%
97.5
≥97.5%
C7H5NO5
183.12

Specifications

3-Hydroxy-4-nitrobenzoic acid
>228°C
98%
97.5
Authentic
Powder
98%
HOC6H3(NO2)CO2H
MFCD00007110
0.5% max.
Yellow to brown
100g
619-14-7
100.0
Plastic bottle
≥97.5%
C7H5NO5
183.12
10, 146
Videos
SDS