missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Maybridge 3-Hydroxy-4-iodobenzaldehyde, 10g, 97%

Brand:  Thermo Scientific Maybridge BTB10430EA

3-Hydroxy-4-iodobenzaldehyde, 10g, 97%

Code : 31

Additional Details : Weight : 0.06000kg

Product Code. 10434142

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.