Learn More
3-Hydroxy-4-iodobenzaldehyde, 97%, Maybridge
Brand: Maybridge BTB10430DA
Code : 31
Additional Details : CAS Number : 13524-71-6 Weight : 0.00100kg
Quantity | 1g |
---|---|
Packaging | Amber Glass Bottle |
Chemical Identifiers
3-Hydroxy-4-iodobenzaldehyde | |
97% | |
248.01 |
97% | |
C7H5IO2 |
Specifications
3-Hydroxy-4-iodobenzaldehyde | |
135242-71-6 | |
97% | |
C7H5IO2 |
1g | |
Amber Glass Bottle | |
97% | |
248.01 |