Learn More
Thermo Scientific Maybridge 3-Hydroxy-4-iodobenzaldehyde, 25g, 97%
Brand: Thermo Scientific Maybridge BTB10430EB
3-Hydroxy-4-iodobenzaldehyde, 25g, 97%
Code : 31
Additional Details : CAS Number : 135242-71-6 Weight : 0.07500kg
Brand: Thermo Scientific Maybridge BTB10430EB
3-Hydroxy-4-iodobenzaldehyde, 25g, 97%
Code : 31
Additional Details : CAS Number : 135242-71-6 Weight : 0.07500kg