missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-quinoxalinecarboxylic Acid 98%, ACROS Organics™

Brand:  Acros Organics 148590050

Code : 29

Additional Details : CAS Number : 1204-75-7 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10552351

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3-Hydroxy-2-quinoxalinecarboxylic acid
1204-75-7
100.0
98%
190.16
98%
97.5
≥97.5%
C9H6N2O3
MFCD00006726

Specifications

3-Hydroxy-2-quinoxalinecarboxylic acid
5g
1204-75-7
100.0
Glass bottle
≥97.5%
C9H6N2O3
MFCD00006726
Green-yellow
98%
97.5
Authentic
Powder and Chunks
98%
190.16
Videos
SDS