3-Hydroxy-2-quinoxalinecarboxylic Acid 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 148590010

1GR 3-Hydroxy-2-quinoxalinecarboxylic acid, 98%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 1204-75-7 Weight: 0.05100kg


Packaging Glass bottle
Quantity 1g

Product Code. 10143050

Quantity Price
1 £ 23.3 / 1g
Estimated Shipment date
from Supplier 03-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-2-quinoxalinecarboxylic acid
Name Note 98%
CAS 1204-75-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H6N2O3
Formula Weight 190.16
MDL Number MFCD00006726

Specification

Assay ≥97.5%
Physical Form Powder and Chunks
Formula Weight 190.16
Quantity 1g
Color Green-yellow
CAS Min % 97.5
EINECS Number 214-877-4
CAS 1204-75-7
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00006726
Packaging Glass bottle
Name Note 98%
Molecular Formula C9H6N2O3
CAS Max % 100.0
Chemical Name or Material 3-Hydroxy-2-quinoxalinecarboxylic acid
TSCA TSCA
Documentation

Technical documents