missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-nitropyridine 97%, ACROS Organics™

Brand:  Acros Organics 148100250

Code : 29

Additional Details : CAS Number : 15128-82-2 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10581771

  • £38.80 / 25g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

3-Hydroxy-2-nitropyridine
15128-82-2
100.0
97%
HO(C5H3N)NO2
MFCD00006258
97%
96.0
≥96.0%
C5H4N2O3
140.1
2-Nitro-3-pyridinol

Specifications

3-Hydroxy-2-nitropyridine
67° to 72°C
97%
96.0
Authentic
Crystalline Powder
97%
HO(C5H3N)NO2
MFCD00006258
Yellow
25g
15128-82-2
100.0
Glass bottle
≥96.0%
C5H4N2O3
140.1
2-Nitro-3-pyridinol
Videos
SDS