missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyne, 99%, Thermo Scientific Chemicals

Catalog Number 10753465
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
928-49-4
C6H10
82.15
MFCD00009381
DQQNMIPXXNPGCV-UHFFFAOYSA-N
3-hexyne, diethylacetylene, unii-9gtq990q4k, acmc-209rhw, c2h5c.$.cc2h5
13568
hex-3-yne
CCC#CCC
This item is not returnable. View return policy
928-49-4
C6H10
82.15
MFCD00009381
DQQNMIPXXNPGCV-UHFFFAOYSA-N
3-hexyne, diethylacetylene, unii-9gtq990q4k, acmc-209rhw, c2h5c.$.cc2h5
13568
hex-3-yne
CCC#CCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7200g/mL
Boiling Point 81°C to 82°C
Flash Point -14°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH2C∽CCH2CH3
Refractive Index 1.4100 to 1.412
Quantity 5 g
Specific Gravity 0.72
Formula Weight 82.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Hexyne, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'