missing translation for 'onlineSavingsMsg'
Learn More

3-Heptanone, 98%, Thermo Scientific Chemicals

Catalog Number 10675922
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
106-35-4
C7H14O
114.19
MFCD00009483
NGAZZOYFWWSOGK-UHFFFAOYSA-N
3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
7802
CHEBI:50139
heptan-3-one
CCCCC(=O)CC
This item is not returnable. View return policy
106-35-4
C7H14O
114.19
MFCD00009483
NGAZZOYFWWSOGK-UHFFFAOYSA-N
3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
7802
CHEBI:50139
heptan-3-one
CCCCC(=O)CC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -39.0°C
Color Yellow
Density 0.8100g/mL
Boiling Point 146.0°C to 149.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)3COC2H5
Refractive Index 1.4075 to 1.4095
Quantity 250 mL
Beilstein 01,699
Specific Gravity 0.81
Solubility Information Solubility in water: 3.3g/L water (20° c). Other solubilities: soluble in alcohol and ether
Formula Weight 114.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Heptanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Heptanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'