missing translation for 'onlineSavingsMsg'
Learn More

3-Furanmethanol, 99%, Thermo Scientific Chemicals

Catalog Number p-3527054
98.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4412-91-3
C5H6O2
98.10
MFCD00005352
STJIISDMSMJQQK-UHFFFAOYSA-N
3-furanmethanol, furan-3-methanol, 3-furylmethanol, 3-hydroxymethyl furan, 3-furfuryl alcohol, 3-hydroxymethylfuran, 3-furyl alcohol, furan-3-yl-methanol, furan-3-yl methanol, 3-hydroxymethyl-furan
20449
furan-3-ylmethanol
OCC1=COC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4412-91-3
C5H6O2
98.10
MFCD00005352
STJIISDMSMJQQK-UHFFFAOYSA-N
3-furanmethanol, furan-3-methanol, 3-furylmethanol, 3-hydroxymethyl furan, 3-furfuryl alcohol, 3-hydroxymethylfuran, 3-furyl alcohol, furan-3-yl-methanol, furan-3-yl methanol, 3-hydroxymethyl-furan
20449
furan-3-ylmethanol
OCC1=COC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1390g/mL
Boiling Point 79.0°C to 80.0°C (17.0 mmHg)
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4835 to 1.4855
Quantity 5 g
Beilstein 17, III, 1266
Specific Gravity 1.139
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 98.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Furanmethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Furanmethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'