missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Furaldehyde, 97%, stab. with ≈0.05% BHT

Brand:  Alfa Aesar™ A11334.14

Code : NEW

Additional Details : CAS Number : 498-60-2 Weight : 0.02500kg Transport : UN number : 1199 Chem class : 1 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 11403744

  • £171.72 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
3-Furaldehyde is compound of honey of various floral origin. It is used to produce N-furan-3-ylmethylene-N′-pyridin-2-yl-hydrazine by reaction with 2-hydrazino-pyridine. It is also used as a solvent.

Solubility
Soluble in water, benzene, chloroform, alcohol and ether.

Notes
Air & Light Sensitive. Store at 4°C. Store in the dark. Incompatible with bases, light and air.
Specifications

Chemical Identifiers

3-Furaldehyde
Stabilized with ≈0.05% BHT
96.09
UN1199
Furan-3-carboxaldehyde
498-60-2
C5H4O2
MFCD00010424
105852

Specifications

3-Furaldehyde
498-60-2
C5H4O2
MFCD00010424
105852
145°C
48°C (118°F)
Stabilized with ≈0.05% BHT
97%
96.09
UN1199
Furan-3-carboxaldehyde
25g
Almond-like
Videos
SDS