missing translation for 'onlineSavingsMsg'
Learn More

3-Formylsalicylic acid hydrate, 97%, Thermo Scientific Chemicals

Catalog Number p-3758054
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1266615-78-4
C8H6O4
166.13
MFCD03804454
YOEUNKPREOJHBW-UHFFFAOYSA-N
3-Formyl-2-hydroxybenzoic acid hydrate, 2-Hydroxyisophthalaldehyde acid hydrate
3-formyl-2-hydroxybenzoic acid
OC(=O)C1=CC=CC(C=O)=C1O
This item is not returnable. View return policy
1266615-78-4
C8H6O4
166.13
MFCD03804454
YOEUNKPREOJHBW-UHFFFAOYSA-N
3-Formyl-2-hydroxybenzoic acid hydrate, 2-Hydroxyisophthalaldehyde acid hydrate
3-formyl-2-hydroxybenzoic acid
OC(=O)C1=CC=CC(C=O)=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 249°C to 252°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 166.13
Percent Purity 97%
Chemical Name or Material 3-Formylsalicylic acid hydrate
compliance-icons compliance-icons
Product Identifier
  • 3-Formylsalicylic acid hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'