missing translation for 'onlineSavingsMsg'
Learn More

3-Formylphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10606354
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
87199-16-4
C7H7BO3
149.94
MFCD00161356
HJBGZJMKTOMQRR-UHFFFAOYSA-N
3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
2734356
(3-formylphenyl)boronic acid
OB(O)C1=CC=CC(C=O)=C1
This item is not returnable. View return policy
87199-16-4
C7H7BO3
149.94
MFCD00161356
HJBGZJMKTOMQRR-UHFFFAOYSA-N
3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
2734356
(3-formylphenyl)boronic acid
OB(O)C1=CC=CC(C=O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 100°C
Color Beige to White or Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 149.94
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 3-Formylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Formylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'