missing translation for 'onlineSavingsMsg'
Learn More

3-Formylbenzonitrile, 97%, Thermo Scientific™

Catalog Number p-2700103
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
24964-64-5
C8H5NO
131.134
MFCD00003344
HGZJJKZPPMFIBU-UHFFFAOYSA-N
3-cyanobenzaldehyde, m-cyanobenzaldehyde, benzonitrile, 3-formyl, m-formylbenzonitrile, m-cyanobenazldehyde, 3-cyano benzaldehyde, 3-formyl-benzonitrile, 3-formylbenzenecarbonitrile, 3cyano-benzaldehyde, m-cyano benzaldehyde
90670
3-formylbenzonitrile
C1=CC(=CC(=C1)C=O)C#N
This item is not returnable. View return policy
24964-64-5
C8H5NO
131.134
MFCD00003344
HGZJJKZPPMFIBU-UHFFFAOYSA-N
3-cyanobenzaldehyde, m-cyanobenzaldehyde, benzonitrile, 3-formyl, m-formylbenzonitrile, m-cyanobenazldehyde, 3-cyano benzaldehyde, 3-formyl-benzonitrile, 3-formylbenzenecarbonitrile, 3cyano-benzaldehyde, m-cyano benzaldehyde
90670
3-formylbenzonitrile
C1=CC(=CC(=C1)C=O)C#N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula NCC6H4CHO
Quantity 1 g
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material 3-Formylbenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'