missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluorophenylglyoxal hydrate, 98%, dry wt. basis

Brand:  Alfa Aesar™ L20278.06

Code : NEW

Additional Details : CAS Number : 121247-01-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11379146

  • £239.20 / 5g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Fluorophenylglyoxal hydrate
C8H5FO2·xH2O
MFCD05664101
121247-01-6
152.13 (Anhydrous)
3537863

Specifications

3-Fluorophenylglyoxal hydrate
5g
98%
C8H5FO2·xH2O
MFCD05664101
66° to 68°C
121247-01-6
(dry wt.)
152.13 (Anhydrous)
3537863
Videos
SDS