missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylacetone, 98%, ACROS Organics™

£26.32 - £96.56

Chemical Identifiers

CAS 1737-19-5
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00061144
InChI Key UWCPYXSRCQVABG-UHFFFAOYSA-N
Synonym 3-fluorophenylacetone, 1-3-fluorophenyl propan-2-one, 1-3-fluorophenyl acetone, acmc-1bqff, m-fluorophenyl-2-propanone, 1-3-fluorophenyl acetone #, 1-3-fluorophenyl-2-propanone
PubChem CID 137185
IUPAC Name 1-(3-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC(=CC=C1)F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10008731
SDS View Documents
Acros Organics
350960010
1g Glass bottle
£26.32
1g
View alternative products
Save Up to 
»
Estimated Shipment
06-05-2021
Please to purchase this item. Need a web account? Register with us today!
10561472
SDS View Documents
Acros Organics
350960050
5g Glass bottle
£96.56
5g
View alternative products
Save Up to 
»
Estimated Shipment
06-05-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1737-19-5
152.17
UWCPYXSRCQVABG-UHFFFAOYSA-N
137185
CC(=O)CC1=CC(=CC=C1)F
C9H9FO
MFCD00061144
3-fluorophenylacetone, 1-3-fluorophenyl propan-2-one, 1-3-fluorophenyl acetone, acmc-1bqff, m-fluorophenyl-2-propanone, 1-3-fluorophenyl acetone #, 1-3-fluorophenyl-2-propanone
1-(3-fluorophenyl)propan-2-one

Specifications

3-Fluorophenylacetone
Glass bottle
Green-Yellow
98%
97.5
97.5% min. (GC)
MFCD00061144
UWCPYXSRCQVABG-UHFFFAOYSA-N
1-(3-fluorophenyl)propan-2-one
137185
Liquid
Authentic
1.4955 to 1.4975 (20°C, 589nm)
1g
1737-19-5
100.0
C9H9FO
3-fluorophenylacetone, 1-3-fluorophenyl propan-2-one, 1-3-fluorophenyl acetone, acmc-1bqff, m-fluorophenyl-2-propanone, 1-3-fluorophenyl acetone #, 1-3-fluorophenyl-2-propanone
CC(=O)CC1=CC(=CC=C1)F
152.17
152.17
98%
SDS