missing translation for 'onlineSavingsMsg'
Learn More

(3-Fluorophenyl)methanesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-3522364
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
24974-72-9
C7H6ClFO2S
208.631
MFCD04117456
QKGIPEQAFOXGSI-UHFFFAOYSA-N
3-fluorophenyl methanesulfonyl chloride, 3-fluoro-phenyl-methanesulfonyl chloride, 3-fluorophenyl methanesulphonyl chloride, benzenemethanesulfonyl chloride, 3-fluoro, chloro 3-fluorophenyl methyl sulfone, pubchem10052, 3-fluorobenzylsulphonyl chloride, benzenemethanesulfonylchloride,3-fluoro
2759107
(3-fluorophenyl)methanesulfonyl chloride
C1=CC(=CC(=C1)F)CS(=O)(=O)Cl
This item is not returnable. View return policy
24974-72-9
C7H6ClFO2S
208.631
MFCD04117456
QKGIPEQAFOXGSI-UHFFFAOYSA-N
3-fluorophenyl methanesulfonyl chloride, 3-fluoro-phenyl-methanesulfonyl chloride, 3-fluorophenyl methanesulphonyl chloride, benzenemethanesulfonyl chloride, 3-fluoro, chloro 3-fluorophenyl methyl sulfone, pubchem10052, 3-fluorobenzylsulphonyl chloride, benzenemethanesulfonylchloride,3-fluoro
2759107
(3-fluorophenyl)methanesulfonyl chloride
C1=CC(=CC(=C1)F)CS(=O)(=O)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material (3-Fluorophenyl)methanesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'