missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenol, 98%, Thermo Scientific Chemicals

Catalog Number 10500931
Click to view available options
Quantity:
10 mL
50 mL
200 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
372-20-3
C6H5FO
112.1
MFCD00002254
SJTBRFHBXDZMPS-UHFFFAOYSA-N
m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
9743
3-fluorophenol
C1=CC(=CC(=C1)F)O
This item is not returnable. View return policy
372-20-3
C6H5FO
112.1
MFCD00002254
SJTBRFHBXDZMPS-UHFFFAOYSA-N
m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
9743
3-fluorophenol
C1=CC(=CC(=C1)F)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 10.0°C
Color Brown-Yellow to Colorless
Density 1.2400g/mL
Boiling Point 178.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4OH
Refractive Index 1.5125 to 1.5155
Quantity 10 mL
Beilstein 06, I, 97
Specific Gravity 1.24
Solubility Information Solubility in water: insoluble
Formula Weight 112.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluorophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'