missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluorobenzylspiperone maleate, Tocris Bioscience™

D2-like receptor ligand

Brand:  Tocris (Bio-Techne) 070110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13487873

  • £128.25 / 10mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

A very potent and selective ligand for the D2 receptor (Ki = 0.023 nM). Compared with spiperone, displays a 2.5-fold greater affinity at D2 and a 12-fold lower affinity at 5-HT2 receptors.
Specifications

Chemical Identifiers

1135278-61-3
619.666
8-[4-(4-Fluorophenyl)-4-oxobutyl]-(3-fluorobenzyl-1-phenyl)-1,3,8-triazaspiro[4,5]decan-4-one maleate
(Z)-but-2-enedioic acid;3-[(3-fluorophenyl)methyl]-8-[4-(4-fluorophenyl)-4-oxobutyl]-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one
C34H35F2N3O6
JURUFADWVGXLOY-BTJKTKAUSA-N
24978532
C1CN(CCC12C(=O)N(CN2C3=CC=CC=C3)CC4=CC(=CC=C4)F)CCCC(=O)C5=CC=C(C=C5)F.C(=CC(=O)O)C(=O)O

Specifications

3'-Fluorobenzylspiperone maleate
1135278-61-3
C34H35F2N3O6
JURUFADWVGXLOY-BTJKTKAUSA-N
(Z)-but-2-enedioic acid;3-[(3-fluorophenyl)methyl]-8-[4-(4-fluorophenyl)-4-oxobutyl]-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one
24978532
10mg
>98%
8-[4-(4-Fluorophenyl)-4-oxobutyl]-(3-fluorobenzyl-1-phenyl)-1,3,8-triazaspiro[4,5]decan-4-one maleate
C1CN(CCC12C(=O)N(CN2C3=CC=CC=C3)CC4=CC(=CC=C4)F)CCCC(=O)C5=CC=C(C=C5)F.C(=CC(=O)O)C(=O)O
619.666
619.67
Videos