missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-2700086
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
100-82-3
C7H8FN
125.15
MFCD00008113
QVSVMNXRLWSNGS-UHFFFAOYSA-N
3-fluorobenzylamine, 3-fluorophenyl methanamine, m-fluorobenzylamine, benzenemethanamine, 3-fluoro, 1-3-fluorophenyl methanamine, 3-fluoro-benzylamine, benzylamine, m-fluoro, 3-fluorophenyl methylamine, m-fluorobenzyl amine, 3-fluorobenzyl amine
66853
(3-fluorophenyl)methanamine
C1=CC(=CC(=C1)F)CN
This item is not returnable. View return policy
100-82-3
C7H8FN
125.15
MFCD00008113
QVSVMNXRLWSNGS-UHFFFAOYSA-N
3-fluorobenzylamine, 3-fluorophenyl methanamine, m-fluorobenzylamine, benzenemethanamine, 3-fluoro, 1-3-fluorophenyl methanamine, 3-fluoro-benzylamine, benzylamine, m-fluoro, 3-fluorophenyl methylamine, m-fluorobenzyl amine, 3-fluorobenzyl amine
66853
(3-fluorophenyl)methanamine
C1=CC(=CC(=C1)F)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 87.0°C to 89.0°C (25.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2NH2
Refractive Index 1.5152
Quantity 1 g
Specific Gravity 1.09
Formula Weight 125.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'