missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl alcohol, 98%, Thermo Scientific™

Catalog Number p-134167
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
456-47-3
C7H7FO
126.13
MFCD00004631
QDHRSLFSDGCJFX-UHFFFAOYSA-N
3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486
68008
(3-fluorophenyl)methanol
C1=CC(=CC(=C1)F)CO
This item is not returnable. View return policy
456-47-3
C7H7FO
126.13
MFCD00004631
QDHRSLFSDGCJFX-UHFFFAOYSA-N
3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486
68008
(3-fluorophenyl)methanol
C1=CC(=CC(=C1)F)CO
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1600g/mL
Boiling Point 104°C to 105°C (22.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2OH
Refractive Index 1.5080 to 1.511
Quantity 5 g
Specific Gravity 1.16
Formula Weight 126.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzyl alcohol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'