missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzenesulfonamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 1524-40-9 |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 131.1°C to 132.7°C |
Color | White to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass Bottle |
Molecular Formula | C6H6FNO2S |
Linear Formula | FC6H4SO2NH2 |
Show More |
Safety and Handling
Product Identifier
- 3-Fluorobenzenesulfonamide
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'