missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzenesulfonamide, 97%, Thermo Scientific™

Catalog Number 15402078
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1524-40-9
C6H6FNO2S
175.177
MFCD00042284
CRINBBOGNYCAOV-UHFFFAOYSA-N
3-fluorobenzene-1-sulfonamide, benzenesulfonamide, 3-fluoro, 3-fluorobenzenesulphonamide, 3-fbsa, 2weo, pubchem11762, acmc-20a5kf, 3-fluoranylbenzenesulfonamide, ksc180k2p, 3-fluorobenzenesulfonamide
150924
3-fluorobenzenesulfonamide
C1=CC(=CC(=C1)S(=O)(=O)N)F
This item is not returnable. View return policy
1524-40-9
C6H6FNO2S
175.177
MFCD00042284
CRINBBOGNYCAOV-UHFFFAOYSA-N
3-fluorobenzene-1-sulfonamide, benzenesulfonamide, 3-fluoro, 3-fluorobenzenesulphonamide, 3-fbsa, 2weo, pubchem11762, acmc-20a5kf, 3-fluoranylbenzenesulfonamide, ksc180k2p, 3-fluorobenzenesulfonamide
150924
3-fluorobenzenesulfonamide
C1=CC(=CC(=C1)S(=O)(=O)N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 124°C to 129°C
Color White to Brown
Quantity 250 mg
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 175.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Fluorobenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'