missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number 10738645
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
701-27-9
C6H4ClFO2S
194.60
MFCD00042286
OKYSUJVCDXZGKE-UHFFFAOYSA-N
3-fluorobenzene-1-sulfonyl chloride, m-fluorobenzenesulfonyl chloride, 3-fluorobenzenesulfonylchloride, 3-fluorobenzenesulphonyl chloride, 3-fluoro-benzenesulfonyl chloride, 3-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 3-fluoro, pubchem5156, acmc-209od0, 3-fluorobenzensulfonylchloride
2734258
3-fluorobenzenesulfonyl chloride
FC1=CC(=CC=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
701-27-9
C6H4ClFO2S
194.60
MFCD00042286
OKYSUJVCDXZGKE-UHFFFAOYSA-N
3-fluorobenzene-1-sulfonyl chloride, m-fluorobenzenesulfonyl chloride, 3-fluorobenzenesulfonylchloride, 3-fluorobenzenesulphonyl chloride, 3-fluoro-benzenesulfonyl chloride, 3-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 3-fluoro, pubchem5156, acmc-209od0, 3-fluorobenzensulfonylchloride
2734258
3-fluorobenzenesulfonyl chloride
FC1=CC(=CC=C1)S(Cl)(=O)=O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula FC6H4SO2Cl
Quantity 1 g
Formula Weight 194.61
Percent Purity 97%
Chemical Name or Material 3-Fluorobenzene-1-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'