missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzene-1-sulfonyl chloride, 97%, Maybridge

Brand:  Maybridge TL00519EB

Code : 31

Additional Details : CAS Number : 701-27-9 Weight : 0.07500kg Transport : UN number : 3265 Chem class : 8 Pack group : III

Quantity 25g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10400594

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3-Fluorobenzene-1-sulfonyl chloride
97.0
97%
FC6H4SO2Cl
MFCD00042286
701-27-9
97%
C6H4ClFO2S
194.61

Specifications

3-Fluorobenzene-1-sulfonyl chloride
701-27-9
Amber Glass Bottle
97%
FC6H4SO2Cl
MFCD00042286
25g
97.0
97%
C6H4ClFO2S
194.61
Videos
SDS