missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3'-Fluoroacetophenone, 98+%

Brand:  Alfa Aesar™ A11733.22

Code : NEW

Additional Details : CAS Number : 455-36-7 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11419263

  • £346.70 / 100g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Description

Description

Applications
3′-Fluoroacetophenone is used as raw material and intermediate in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents. Ensure adequate ventilation during use. Use only in a chemical fume hood.
Specifications

Chemical Identifiers

3'-Fluoroacetophenone
C8H7FO
MFCD00000338
455-36-7
138.14
907041

Specifications

3'-Fluoroacetophenone
80° to 81°C (9mmHg)
100g
≥98%
138.14
907041
80°C (176°F)
−3°C
455-36-7
C8H7FO
MFCD00000338
Videos
SDS