missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)phenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4228694
99.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
159020-59-4
C7H5BF4O2
207.92
MFCD06657735
WEMCWZGCSRGJGW-UHFFFAOYSA-N
3-fluoro-5-trifluoromethyl phenyl boronic acid, 3-fluoro-5-trifluoromethyl phenylboronic acid, 3-fluoro-5-trifluoromethylphenylboronic acid, 3-borono-5-fluorobenzotrifluoride, 3-fluoro-5-trifluoromethyl benzeneboronic acid, 3-fluoro-5-trifluoromethylphenyl boronic acid, 3-trifluoromethyl-5-fluoro-phenylboronic acid, 3-fluoro-5-trifluoromethyl benzene boronic acid, 5-fluoro-3-trifluoromethylbenzeneboronic acid
2783192
[3-fluoro-5-(trifluoromethyl)phenyl]boronic acid
OB(O)C1=CC(F)=CC(=C1)C(F)(F)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
159020-59-4
C7H5BF4O2
207.92
MFCD06657735
WEMCWZGCSRGJGW-UHFFFAOYSA-N
3-fluoro-5-trifluoromethyl phenyl boronic acid, 3-fluoro-5-trifluoromethyl phenylboronic acid, 3-fluoro-5-trifluoromethylphenylboronic acid, 3-borono-5-fluorobenzotrifluoride, 3-fluoro-5-trifluoromethyl benzeneboronic acid, 3-fluoro-5-trifluoromethylphenyl boronic acid, 3-trifluoromethyl-5-fluoro-phenylboronic acid, 3-fluoro-5-trifluoromethyl benzene boronic acid, 5-fluoro-3-trifluoromethylbenzeneboronic acid
2783192
[3-fluoro-5-(trifluoromethyl)phenyl]boronic acid
OB(O)C1=CC(F)=CC(=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 178.0°C to 180.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 207.92
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • (3-Fluoro-5-trifluoromethylphenyl)boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'