missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-5-(trifluoromethoxy)benzamide, 97%

£33.00 - £109.00

Chemical Identifiers

CAS 1352999-12-2
Molecular Formula C8H5F4NO2
Molecular Weight (g/mol) 223.127
MDL Number MFCD18783223
InChI Key UWJVDWZVDASOBE-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethoxy benzamide
PubChem CID 66523540
IUPAC Name 3-fluoro-5-(trifluoromethoxy)benzamide
SMILES C1=C(C=C(C=C1OC(F)(F)F)F)C(=O)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15437217 SDS View Documents Alfa Aesar™
H34282.MD
250mg £33.00
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 05-03-2020
Please to purchase this item. Need a web account? Register with us today!
15427217 SDS View Documents Alfa Aesar™
H34282.03
1g £109.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 05-03-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

1352999-12-2
223.127
UWJVDWZVDASOBE-UHFFFAOYSA-N
66523540
C1=C(C=C(C=C1OC(F)(F)F)F)C(=O)N
C8H5F4NO2
MFCD18783223
3-fluoro-5-trifluoromethoxy benzamide
3-fluoro-5-(trifluoromethoxy)benzamide

Specifications

3-Fluoro-5-(trifluoromethoxy)benzamide
1352999-12-2
MFCD18783223
UWJVDWZVDASOBE-UHFFFAOYSA-N
3-fluoro-5-(trifluoromethoxy)benzamide
66523540
97%
250mg
C8H5F4NO2
3-fluoro-5-trifluoromethoxy benzamide
C1=C(C=C(C=C1OC(F)(F)F)F)C(=O)N
223.127
223.12