missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number 15477686
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
710348-94-0
C12H15BFNO4
267.063
CKBJFSWEKLWHTI-UHFFFAOYSA-N
2-3-fluoro-5-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-fluoro-5-nitrophenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
59770452
2-(3-fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)[N+](=O)[O-]
This item is not returnable. View return policy
710348-94-0
C12H15BFNO4
267.063
CKBJFSWEKLWHTI-UHFFFAOYSA-N
2-3-fluoro-5-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-fluoro-5-nitrophenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
59770452
2-(3-fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 267.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-5-nitrobenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'