missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15427906
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1416367-00-4
C13H18BFO3
252.09
MFCD16996298
DGHRKRRFVKPHDQ-UHFFFAOYSA-N
2-3-fluoro-5-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-5-methoxyphenylboronic acid pinacol ester, 3-fluoro-5-methoxybenzeneboronic acid pinacol ester
72221149
2-(3-fluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
COC1=CC(F)=CC(=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
1416367-00-4
C13H18BFO3
252.09
MFCD16996298
DGHRKRRFVKPHDQ-UHFFFAOYSA-N
2-3-fluoro-5-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-5-methoxyphenylboronic acid pinacol ester, 3-fluoro-5-methoxybenzeneboronic acid pinacol ester
72221149
2-(3-fluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
COC1=CC(F)=CC(=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 252.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-5-methoxybenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'