missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-5-formylbenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62303.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417266

  • £302.30 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Fluoro-5-formylbenzeneboronic acid pinacol ester
250.07
3-Fluoro-5-formylphenylboronic acid pinacol ester

Specifications

3-Fluoro-5-formylbenzeneboronic acid pinacol ester
1112209-24-1
96%
3-Fluoro-5-formylphenylboronic acid pinacol ester
1g
Solid
250.07
Videos
SDS