missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-5-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62773.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487826

  • £352.70 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Fluoro-5-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
321.19
3-Fluoro-5-(4-morpholinylmethyl)phenylboronic acid pinacol ester

Specifications

3-Fluoro-5-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
Solid
321.19
1g
96%
3-Fluoro-5-(4-morpholinylmethyl)phenylboronic acid pinacol ester
Videos
SDS