missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylpyridine, 95%, Thermo Scientific™

Catalog Number 15432069
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
399-88-2
C6H6FN
111.119
MFCD04114226
WZMOEPZZTTWDIA-UHFFFAOYSA-N
3-fluoro-4-picoline, 4-methyl-3-fluoropyridine, pyridine, 3-fluoro-4-methyl, zlchem 994, pubchem9251, acmc-1agea, 3-fluoro-4-methyl pyridine, 3-fluoro-4-methyl-pyridine, 3-fluoro-4-methylpyridine?, ksc925s4f
345367
3-fluoro-4-methylpyridine
CC1=C(C=NC=C1)F
This item is not returnable. View return policy
399-88-2
C6H6FN
111.119
MFCD04114226
WZMOEPZZTTWDIA-UHFFFAOYSA-N
3-fluoro-4-picoline, 4-methyl-3-fluoropyridine, pyridine, 3-fluoro-4-methyl, zlchem 994, pubchem9251, acmc-1agea, 3-fluoro-4-methyl pyridine, 3-fluoro-4-methyl-pyridine, 3-fluoro-4-methylpyridine?, ksc925s4f
345367
3-fluoro-4-methylpyridine
CC1=C(C=NC=C1)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.08 g/mL
Boiling Point 136°C (748 mmHg)
Refractive Index 1.4795
Quantity 1 g
UN Number UN1993
Beilstein 107788
Formula Weight 111.12
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Fluoro-4-methylpyridine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'