missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzyl bromide, 97%, Thermo Scientific™

Catalog Number p-7028932
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
145075-44-1
C8H8BrF
203.05
MFCD01631534
YOACSNDETIDTAY-UHFFFAOYSA-N
3-fluoro-4-methylbenzyl bromide, 4-bromomethyl-2-fluoro-1-methylbenzene, benzene, 4-bromomethyl-2-fluoro-1-methyl, acmc-1c02p, 3-fluoro-4-methyl-benzyl bromide, 3-fluoro-4-methylbenzylbromide, benzene,4-bromomethyl-2-fluoro-1-methyl, 4-bromomethyl-2-fluoranyl-1-methyl-benzene
2774603
4-(bromomethyl)-2-fluoro-1-methylbenzene
CC1=C(F)C=C(CBr)C=C1
This item is not returnable. View return policy
145075-44-1
C8H8BrF
203.05
MFCD01631534
YOACSNDETIDTAY-UHFFFAOYSA-N
3-fluoro-4-methylbenzyl bromide, 4-bromomethyl-2-fluoro-1-methylbenzene, benzene, 4-bromomethyl-2-fluoro-1-methyl, acmc-1c02p, 3-fluoro-4-methyl-benzyl bromide, 3-fluoro-4-methylbenzylbromide, benzene,4-bromomethyl-2-fluoro-1-methyl, 4-bromomethyl-2-fluoranyl-1-methyl-benzene
2774603
4-(bromomethyl)-2-fluoro-1-methylbenzene
CC1=C(F)C=C(CBr)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 77°C (3 mmHg)
Refractive Index 1.546
Quantity 1 g
UN Number UN3265
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylbenzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'