missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7054603
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1415960-53-0
C15H20BFO4
294.129
MMANYAVWRYOHTI-UHFFFAOYSA-N
3-fluoro-4-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 3-fluoro-4-methoxycarbonylmethyl benzeneboronicacidpinacolester, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetic acid methyl ester
71265929
methyl 2-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CC(=O)OC)F
This item is not returnable. View return policy
1415960-53-0
C15H20BFO4
294.129
MMANYAVWRYOHTI-UHFFFAOYSA-N
3-fluoro-4-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 3-fluoro-4-methoxycarbonylmethyl benzeneboronicacidpinacolester, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetic acid methyl ester
71265929
methyl 2-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CC(=O)OC)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 294.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'