missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62503.03

Code : NEW

Additional Details : CAS Number : 603122-52-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15427506

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester
280.1
3-Fluoro-4-(methoxycarbonyl)phenylboronic acid pinacol ester
603122-52-7
MFCD16996314

Specifications

3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester
603122-52-7
96%
MFCD16996314
1g
Solid
280.1
3-Fluoro-4-(methoxycarbonyl)phenylboronic acid pinacol ester
Videos
SDS