missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-4-iodobenzoic acid, 98%

Brand:  Alfa Aesar™ H61724.06

Code : NEW

Additional Details : CAS Number : 825-98-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15468288

  • £298.92 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Fluoro-4-iodobenzoic acid
825-98-9
266.01
MFCD07774250

Specifications

3-Fluoro-4-iodobenzoic acid
5g
Solid
266.01
Light Sensitive
825-98-9
98%
MFCD07774250
Videos
SDS