missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hydroxyphenylacetic acid, 98%, Thermo Scientific™

Catalog Number p-4089470
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
458-09-3
C8H7FO3
170.139
MFCD00004348
YRFBZAHYMOSSGX-UHFFFAOYSA-N
3-fluoro-4-hydroxyphenylacetic acid, 2-3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxybenzeneacetic acid, benzeneacetic acid, 3-fluoro-4-hydroxy, 2-3-fluoro-4-hydroxy-phenyl acetic acid, 4-carboxymethyl-2-fluorophenol, 3-fluoro-4-hydroxy-phenyl-acetic acid, acmc-1ahjh, opera_id_1839
68014
2-(3-fluoro-4-hydroxyphenyl)acetic acid
C1=CC(=C(C=C1CC(=O)O)F)O
This item is not returnable. View return policy
458-09-3
C8H7FO3
170.139
MFCD00004348
YRFBZAHYMOSSGX-UHFFFAOYSA-N
3-fluoro-4-hydroxyphenylacetic acid, 2-3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxybenzeneacetic acid, benzeneacetic acid, 3-fluoro-4-hydroxy, 2-3-fluoro-4-hydroxy-phenyl acetic acid, 4-carboxymethyl-2-fluorophenol, 3-fluoro-4-hydroxy-phenyl-acetic acid, acmc-1ahjh, opera_id_1839
68014
2-(3-fluoro-4-hydroxyphenyl)acetic acid
C1=CC(=C(C=C1CC(=O)O)F)O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 131°C to 135°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H3(OH)CH2CO2H
Quantity 1 g
Beilstein 10, III, 440
Merck Index 10, 4079
Formula Weight 170.14
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-hydroxyphenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluoro-4-hydroxyphenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'